Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-SOX2 Rabbit Antibody
  BSBTPA2284
undefined
Anti-SOX2 Rabbit Antibody
  BSBTPA2284

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    SRY (sex determining region Y)-box 2
  • Antigen-Symbol:
    SOX2
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Kreuzadsorption:
    No
  • Form:
    Lyophilized
  • Antigen-Synonyme:
    MCOPS3|ANOP3|SRY-related HMG-box gene 2|transcription factor SOX-2|transcription factor SOX2|sex determining region Y box 2
  • Aminosäurezahl:
    171 to 183
  • UniProtKB:
    P48431
  • Sequence:
    YSMMQDQLGYPQH
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human SOX2 (171-183aa YSMMQDQLGYPQH), different from the related mouse and rat sequence by one amino acid.
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Transcription factor SOX-2(SOX2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

SRY(sex determining region Y)-box 2, also known as SOX2, is a transcription factor that is essential for maintaining self-renewal, or pluripotency of undifferentiated embryonic stem cells. Sox2 is a member of the Sox family of transcription factors, which have been shown to play key roles in many stages of mammalian development. This gene is mapped to 3q26.33. It is found that SOX2 can regulate OCT3/4 expression and maintains ES pluripotency through upstream transcription factors. SOX2 is identified as a lineage-survival oncogene in lung and esophageal squamous cell carcinoma. In addition to those, SOX2 has a critical role in maintenance of embryonic and neural stem cells and holds great promise in research involving induced pluripotency, an emerging and very promising field of regenerative medicine.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SOX2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat