Anti-TLR1 Rabbit Antibody
Catalog # BSBTPA2246
Supplier: Boster Bio
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Toll-like Receptor 1
- Antigen-Symbol:TLR1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Epitop:C-Terminal
- Kreuzadsorption:No
- Form:Lyophilised
- Antigen-Synonyme:rsc786|TLR1|Toll-like receptor 1|TIL. LPRS5|KIAA0012|CD281|TIL
- Aminosäurezahl:771 - 786
- Sequence:NLRAAINIKLTEQAKK
- Lagertemperatur:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TLR1 (771-786aa NLRAAINIKLTEQAKK).
- Reinigung:purified by immunogen affinity purification
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Toll-like receptor 1(TLR1) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: TLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human