Order Entry
Switzerland
ContactUsLinkComponent
Anti-RIP3 Rabbit Antibody
Katalognr. BSBTPA2242
Anbieter:  Boster Bio
undefined
Anti-RIP3 Rabbit Antibody
Katalognr. BSBTPA2242
Anbieter:  Boster Bio

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Receptor-interacting serine/threonine-protein kinase 3
  • Antigen-Symbol:
    RIPK3
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Epitop:
    near the N-terminus
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Antigen-Synonyme:
    RIP-like protein kinase 3|Receptor-interacting protein 3|RIPK3|RIP-3|RIP3|Receptor-interacting serine/threonine-protein kinase 3
  • Aminosäurezahl:
    60 - 76
  • Sequence:
    EVKAMASLDNEFVLRLE
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human RIP3 (60-76aa EVKAMASLDNEFVLRLE).
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    100 µG

Technische Daten

Über diesen Artikel

Rabbit IgG polyclonal antibody for Receptor-interacting serine/threonine-protein kinase 3(RIPK3) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: RIPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human