Order Entry
Switzerland
ContactUsLinkComponent
Anti-RGS9 Rabbit Antibody
Katalognr. BSBTPA2241
Anbieter:  Boster Bio
undefined
Anti-RGS9 Rabbit Antibody
Katalognr. BSBTPA2241
Anbieter:  Boster Bio

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Regulator of G-protein Signaling 9
  • Antigen-Symbol:
    RGS9
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    near the C-terminus
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Antigen-Synonyme:
    MGC26458|RGS9L|MGC111763|PERRS
  • Aminosäurezahl:
    624 - 638
  • Sequence:
    LKSKRVANFFQIKMD
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human RGS9 (624-638aa LKSKRVANFFQIKMD), different from the related mouse and rat sequences by three amino acids.
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    100 µG

Technische Daten

Über diesen Artikel

Rabbit IgG polyclonal antibody for Regulator of G-protein signaling 9(RGS9) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: RGS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat