Order Entry
Switzerland
ContactUsLinkComponent
Anti-PSD95 Rabbit Antibody
N° de catalogue BSBTPA2295
Fournisseur:  Boster Bio
undefined
Anti-PSD95 Rabbit Antibody
N° de catalogue BSBTPA2295
Fournisseur:  Boster Bio

Spécifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Discs Large MAGUK Scaffold Protein 4
  • Antigen-Symbol:
    DLG4
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    near the C-terminus
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Antigen-Synonyme:
    PSD-95 |SAP90|FLJ98574|PSD95|SAP-90 |FLJ97752
  • Aminosäurezahl:
    492 - 507
  • Sequence:
    RRVERREWSRLKAKDW
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human PSD95 (492-507aa RRVERREWSRLKAKDW), identical to the related rat and mouse sequences.
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    100 µG

Spécifications

À propos de cet article

Rabbit IgG polyclonal antibody for Disks large homolog 4(DLG4) detection. Tested with WB, IHC-P in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: DLG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat