Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Anti-EFCAB4A Rabbit Polyclonal Antibody | Avantor

Anti-EFCAB4A Rabbit Polyclonal Antibody

100 µg
Katalognr. USBI146755
Anbieter:   US Biological
Anti-EFCAB4A Rabbit Polyclonal Antibody
Katalognr. USBI146755
Anbieter:  US Biological
Anti-EFCAB4A Rabbit Polyclonal Antibody
Anti-EFCAB4A Rabbit Polyclonal Antibody

100 µg

Preis
SFr. 817.00
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: EFCAB4A (EF-hand calcium binding domain 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Dokumente

Technische Daten
  • Größe:
    100 µg
  • Western blot:
    Yes
  • Antikörper-Typ:
    Primär
  • Konjugation:
    Unconjugated
  • Form:
    liquid
  • Reinigung:
    purified by immunoaffinity chromatography
  • Transporttemperatur:
    Ice
  • Reaktivität:
    Human|Rat|Mouse
  • Konzentration:
    ~1 mg/ml
  • Cat. No.:
    USBI146755
  • Antigen-Name:
    EF-hand calcium binding domain 4A
  • Lagertemperatur:
    Cool
  • Lagerungspuffer:
    PBS, 0,02% sodium azide.
  • Antigen-Symbol:
    EFCAB4A
  • Klonalität:
    Polyklonal
  • Isotyp:
    IgG
  • Wirt:
    Rabbit
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.