Anti-TRAPPC3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI043197-PE
- Antikörper-Typ:Primär
- Antigen-Name:Trafficking Protein Particle Complex 3
- Antigen-Symbol:TRAPPC3
- Klonalität:Polyklonal
- Konjugation:PE (Phycoerythrin)
- ELISA:Yes
- Durchflusszytometrie:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Epitop:N-terminal
- Form:liquid
- Aminosäurezahl:24 to 54
- Lagerungspuffer:PBS, pH 7,2
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Validierte Anwendungen:Fluoresence ELISA
- Reinigung:purified by Protein A chromatography
- Größe:200 µl
- VE:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: TRAPPC3 (trafficking protein particle complex 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human