Anti-EFCAB4B Rabbit Polyclonal Antibody (Biotin)
200 µl
Katalognr. USBI034936-BIOTIN
Anbieter: US Biological
Anti-EFCAB4B Rabbit Polyclonal Antibody (Biotin)
Katalognr. USBI034936-BIOTIN
Anbieter: US Biological
Direkt zu
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: EFCAB4B (EF-hand calcium binding domain 4B)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Dokumente
Technische Daten
- Größe:200 µl
- Western blot:Yes
- Aminosäurezahl:47 to 74
- Antikörper-Typ:Primär
- Konjugation:Biotin
- Form:liquid
- Reinigung:purified by Protein A chromatography
- Transporttemperatur:Ice
- Reaktivität:Human
- Epitop:N-terminal
- Cat. No.:USBI034936-BIOTIN
- Antigen-Name:EF-hand calcium binding domain 4B
- Lagertemperatur:Cool
- Lagerungspuffer:PBS, pH 7,2
- Antigen-Symbol:EFCAB4B
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit
- ELISA:Yes



