Anti-E2F3 Rabbit Polyclonal Antibody
200 µl
Katalognr. USBI034889
Anbieter: US Biological
Direkt zu
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: E2F3 (E2F transcription factor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Dokumente
Technische Daten
- Größe:200 µl
- Western blot:Yes
- Aminosäurezahl:150 to 180
- Antikörper-Typ:Primär
- Konjugation:Unconjugated
- Form:liquid
- Reinigung:purified by Protein A chromatography
- Transporttemperatur:Ice
- Reaktivität:Human
- Epitop:internal
- Cat. No.:USBI034889
- Antigen-Name:E2F transcription factor 3
- Lagertemperatur:Cool
- Lagerungspuffer:PBS, pH 7,2, 0,09% sodium azide.
- Antigen-Symbol:E2F3
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit
- ELISA:Yes



