Order Entry
Switzerland
ContactUsLinkComponent
Anti-CLPTM1L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  USBI034013-FITC
undefined
Anti-CLPTM1L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  USBI034013-FITC

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CLPTM1-like
  • Antigen-Symbol:
    CLPTM1L
  • Klonalität:
    Polyklonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Form:
    liquid
  • Lagerungspuffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by Protein A chromatography
  • Größe:
    200 µl
  • VE:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CLPTM1L (CLPTM1-like)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human