Anti-APPBP2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
USBI031998-AP
- Antikörper-Typ:Primär
- Antigen-Name:Amyloid Beta Precursor Protein (Cytoplasmic Tail) Binding Protein 2
- Antigen-Symbol:APPBP2
- Klonalität:Polyklonal
- Konjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Epitop:internal
- Form:liquid
- Aminosäurezahl:242 to 271
- Lagerungspuffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Reinigung:purified by Protein A chromatography
- Größe:200 µl
- VE:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: APPBP2 (amyloid beta precursor protein (cytoplasmic tail) binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human