Order Entry
Switzerland
ContactUsLinkComponent
Anti-Kallikrein 9 Rabbit Polyclonal Antibody
Katalognr. BSBTPA1820
Anbieter:  Boster Bio
undefined
Anti-Kallikrein 9 Rabbit Polyclonal Antibody
Katalognr. BSBTPA1820
Anbieter:  Boster Bio

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Kallikrein 9
  • Antigen-Symbol:
    KLK9
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Kreuzadsorption:
    No
  • Form:
    Lyophilized
  • Antigen-Synonyme:
    KLK9|KLK-L3|Kallikrein-9 [Precursor]|Kallikrein-like protein 3
  • Aminosäurezahl:
    236 to 250
  • Sequence:
    SVCHYLDWIQEIMEN
  • Lagertemperatur:
    –20 °C
  • Transporttemperatur:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Kallikrein 9 (236-250aa SVCHYLDWIQEIMEN).
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    0,1 mg

Technische Daten

Über diesen Artikel

Rabbit IgG polyclonal antibody for Kallikrein-9(KLK9) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Kallikrein 9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human