Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Anti-EIF4B Rabbit Polyclonal Antibody | Avantor

Anti-EIF4B Rabbit Polyclonal Antibody

100 μg
Katalognr. NOVUNBP2-24475
Anbieter:   Novus Biologicals
Anti-EIF4B Rabbit Polyclonal Antibody
Katalognr. NOVUNBP2-24475
Anbieter:  Novus Biologicals
Anti-EIF4B Rabbit Polyclonal Antibody
Anti-EIF4B Rabbit Polyclonal Antibody

100 μg

Preis
SFr. 510.00
Einloggen für Verfügbarkeit
1 * 0,1 mg
Product Details & Documents

Type: Primary
Antigen: EIF4B (eukaryotic translation initiation factor 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:

Technische Daten
  • Größe:
    100 μg
  • Western blot:
    Yes
  • Cat. No.:
    NOVUNBP2-24475
  • Antigen-Name:
    Eukaryotic translation initiation factor 4B
  • Lagertemperatur:
    Cool
  • Gen-ID:
    1975
  • Antikörper-Typ:
    Primär
  • Konjugation:
    Unconjugated
  • Transporttemperatur:
    Ice
  • Antigen-Synonyme:
    EIF-4B|PRO1843
  • Antigen-Symbol:
    EIF4B
  • Klonalität:
    Polyklonal
  • Wirt:
    Rabbit
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.