Order Entry
Switzerland
ContactUsLinkComponent
Anti-Prosurfactant Protein C Rabbit Polyclonal Antibody
Anti-Prosurfactant Protein C Rabbit Polyclonal Antibody
Codice catalogo BSBTPA1933
Nome del fornitore:  Boster Bio
undefined
Anti-Prosurfactant Protein C Rabbit Polyclonal Antibody
Codice catalogo BSBTPA1933
Nome del fornitore:  Boster Bio

Specifiche

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Prosurfactant Protein C
  • Antigen-Symbol:
    ProSP-C
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    N-Terminal
  • Kreuzadsorption:
    No
  • Form:
    Lyophilized
  • Aminosäurezahl:
    21 to 36
  • Sequence:
    RSQFRIPCCPVHLKRL
  • Lagertemperatur:
    –20 °C
  • Transporttemperatur:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of mouse Prosurfactant Protein C (21-36aa RSQFRIPCCPVHLKRL), identical to the related rat sequence and different from the related human sequence by three amino acids.
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    0,1 mg

Specifiche

Informazioni su questo articolo

Type: Primary
Antigen: Prosurfactant Protein C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat