Anti-TRIF Rabbit Polyclonal Antibody
BSBTPA1902
- Antikörper-Typ:Primär
- Antigen-Name:Toll-like receptor adaptor molecule 1
- Antigen-Symbol:TICAM1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Epitop:C-Terminal
- Kreuzadsorption:No
- Form:Lyophilised
- Antigen-Synonyme:hCG_39321|Trif|HSD34|RNF36
- Aminosäurezahl:692 - 712
- Sequence:NNHMWNQRGSQAPEDKTQEAE
- Lagertemperatur:–20 °C
- Transporttemperatur:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TRIF (692-712aa NNHMWNQRGSQAPEDKTQEAE).
- Reinigung:purified by immunogen affinity purification
- Größe:100 µg
- VE:0,1 mg
Rabbit IgG polyclonal antibody for TIR domain-containing adapter molecule 1 (TICAM1) detection. Tested with WB, IHC-P, IHC-F in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: TRIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human