Order Entry
Switzerland
ContactUsLinkComponent
Anti-TNFAIP1 Rabbit Polyclonal Antibody
Anti-TNFAIP1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1305
Supplier:  Boster Bio
undefined
Anti-TNFAIP1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1305
Supplier:  Boster Bio

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Tumor Necrosis Factor Alpha-induced Protein 1
  • Antigen-Symbol:
    TNFAIP1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    N-Terminal
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Antigen-Synonyme:
    EDP1|hBACURD2|B12|BTBD34|B61
  • Aminosäurezahl:
    27 - 52
  • Sequence:
    NKYVQLNVGGSLYYTTVRALTRHDTM
  • Lagertemperatur:
    –20 °C
  • Transporttemperatur:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human TNFAIP1 (27-52aa NKYVQLNVGGSLYYTTVRALTRHDTM), identical to the related rat and mouse sequences
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2 (TNFAIP1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: TNFAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat