Order Entry
Switzerland
ContactUsLinkComponent
Anti-GFAP Rabbit Polyclonal Antibody
Katalognr. BSBTPA1239
Anbieter:  Boster Bio
undefined
Anti-GFAP Rabbit Polyclonal Antibody
Katalognr. BSBTPA1239
Anbieter:  Boster Bio

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Glial fibrillary acidic protein
  • Antigen-Symbol:
    GFAP
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Antigen-Synonyme:
    FLJ45472|Intermediate filament protein.|Astrocyte|p-GFAP Ser38|GFAP phospho S38|GFAP|Glial Fibrillary Acidic Protein
  • Aminosäurezahl:
    417 - 432
  • Sequence:
    DGEVIKESKQEHKDVM
  • Lagertemperatur:
    –20 °C
  • Transporttemperatur:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human GFAP (417-432aa DGEVIKESKQEHKDVM), identical to the related rat sequence, and different from the related mouse sequence by two amino acids
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    0,1 mg

Technische Daten

Über diesen Artikel

Rabbit IgG polyclonal antibody for Glial fibrillary acidic protein (GFAP) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat