Order Entry
Switzerland
ContactUsLinkComponent
Anti-ATOH8 Rabbit Polyclonal Antibody
  ABCAAB106377-100
 :  Abcam
undefined
Anti-ATOH8 Rabbit Polyclonal Antibody
  ABCAAB106377-100
 :  Abcam

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Protein Atonal homolog 8
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Größe:
    100 µg
  • Packung:
    Vial
  • VE:
    100 µG

 

 

Rabbit polyclonal to ATOH8.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ATOH8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse