Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:alpha thalassemia/mental retardation syndrome X-linked
- Antigen-Symbol:ATRX
- Klonalität:Monoklonal
- Klon:Rad54 4E3/1
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- Wirt:Mouse
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Isotyp:
- Reaktivität:Human
- Western blot:Yes
- Environmentally Preferable:
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µg
- VE:50 µG
Technische Daten
Über diesen Artikel
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATRX (alpha thalassemia/mental retardation syndrome X-linked)
Clonality: monoclonal
Clone: Rad54 4E3/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human