Order Entry
Switzerland
ContactUsLinkComponent
Anti-LAP2 alpha Rabbit Polyclonal Antibody
Anti-LAP2 alpha Rabbit Polyclonal Antibody
  ABCAAB5162-100
 :  Abcam
Anti-LAP2 alpha Rabbit Polyclonal Antibody
  ABCAAB5162-100
 :  Abcam

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    LAP2 alpha
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Immunpräzipitation:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Größe:
    100 µl
  • VE:
    100 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LAP2 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat