Order Entry
Switzerland
ContactUsLinkComponent
Anti-RRAS Mouse Monoclonal Antibody
Catalog # BIRBORB95651-50
Anbieter:  Biorbyt
undefined
Anti-RRAS Mouse Monoclonal Antibody
Catalog # BIRBORB95651-50
Anbieter:  Biorbyt

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    related RAS viral (r-ras) oncogene homolog
  • Antigen-Symbol:
    RRAS
  • Klonalität:
    Monoklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Mouse
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG2b kappa
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Lagerungspuffer:
    HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by ammonium sulfate precipitation
  • Größe:
    50 µg
  • VE:
    50 µG

Technische Daten

Über diesen Artikel

Immunochemistry: 1:400; Western Blot: 1:2000

Type: Primary
Antigen: RRAS (related RAS viral (r-ras) oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human