Anti-ADRM1 Rabbit Polyclonal Antibody
100 µg
Katalognr. BIRBORB77825-100
Marketsource
Anbieter: Biorbyt
Direkt zu
Product Details & Documents
ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000
Type: Primary
Antigen: ADRM1 (adhesion regulating molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Dokumente
orb77825
Technisches Datenblatt
Technische Daten
- Größe:100 µg
- Western blot:Yes
- Antikörper-Typ:Primär
- Konjugation:Unconjugated
- Reinigung:purified by affinity chromatography
- Transporttemperatur:Ice
- Reaktivität:Human
- Cat. No.:BIRBORB77825-100
- Antigen-Name:adhesion regulating molecule 1
- Lagertemperatur:Cool
- Immunogen:Recombinant protein
- Antigen-Symbol:ADRM1
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit
- ELISA:Yes



