Order Entry
Switzerland
ContactUsLinkComponent
Anti-SLC10A1 Rabbit Polyclonal Antibody
Katalognr. BIRBORB76184-100
Anbieter:  Biorbyt
undefined
Anti-SLC10A1 Rabbit Polyclonal Antibody
Katalognr. BIRBORB76184-100
Anbieter:  Biorbyt

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    solute carrier family 10 (sodium/bile acid cotransporter), member 1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Western blot:
    Yes
  • Form:
    lyophilized
  • Aminosäurezahl:
    141 to 154
  • Lagerungspuffer:
    5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
  • Sequence:
    YSRGIYDGDLKDKV
  • Lagertemperatur:
    Cool
  • Konzentration:
    0,1-0,5 ug/ml,0,5-1 ug/ul
  • Transporttemperatur:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human SLC10A1(141-154aa YSRGIYDGDLKDKV), different from the related rat and mouse sequences by one amino acid.
  • Reinigung:
    purified by immunoaffinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

Technische Daten

Über diesen Artikel

Type: Primary
Antigen: SLC10A1 (solute carrier family 10 (sodium/bile acid cotransporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: