Anti-NEK4 Rabbit Polyclonal Antibody
BIRBORB73762-100
: Biorbyt
- Antikörper-Typ:Primär
- Antigen-Name:NIMA-related kinase 4
- Antigen-Symbol:NEK4
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:
- Reaktivität:Human
- Western blot:Yes
- Environmentally Preferable:
- Lagertemperatur:Cool
- Konzentration:0,37 mg/ml
- Transporttemperatur:Ice
- Größe:100 µl
- VE:100 µl
Type: Primary
Antigen: NEK4 (NIMA-related kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human