Anti-RASGRP4 Rabbit Polyclonal Antibody
BIRBORB69878-100
: Biorbyt
- Antikörper-Typ:Primär
- Antigen-Name:RAS guanyl releasing protein 4
- Antigen-Symbol:RASGRP4
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Western blot:Yes
- Lagertemperatur:Cool
- Konzentration:1 mg/ml
- Transporttemperatur:Ice
- Größe:100 µl
- VE:100 µl
Type: Primary
Antigen: RASGRP4 (RAS guanyl releasing protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human