Anti-EVC2 Rabbit Polyclonal Antibody
Catalog # BIRBORB69877-100
Supplier: Biorbyt
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Ellis van Creveld syndrome 2
- Antigen-Symbol:EVC2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Western blot:Yes
- Lagertemperatur:Cool
- Konzentration:1 mg/ml
- Transporttemperatur:Ice
- Größe:100 µl
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: EVC2 (Ellis van Creveld syndrome 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human