Order Entry
Switzerland
ContactUsLinkComponent
Anti-S100A12 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5®)
  BIRBORB129824-100
 :  Biorbyt
Anti-S100A12 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5®)
  BIRBORB129824-100
 :  Biorbyt

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    S100 Calcium Binding Protein A12
  • Antigen-Symbol:
    S100A12
  • Klonalität:
    Polyklonal
  • Konjugation:
    PE (Phycoerythrin)/Cy5®
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Form:
    liquid
  • Lagerungspuffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Immunogen:
    Synthetic peptide
  • Reinigung:
    purified by affinity chromatography
  • Größe:
    100 µl
  • VE:
    100 µl

 

 

Type: Primary
Antigen: S100A12 (S100 calcium binding protein A12)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: