Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Anti-CCR8 Rabbit Polyclonal Antibody

Anti-CCR8 Rabbit Polyclonal Antibody

100 µl
Katalognr. BIRBORB107482-100
Marketsource
Anbieter:   Biorbyt
Anti-CCR8 Rabbit Polyclonal Antibody
Katalognr. BIRBORB107482-100
Anbieter:  Biorbyt
Marketsource
Anti-CCR8 Rabbit Polyclonal Antibody
Anti-CCR8 Rabbit Polyclonal Antibody

100 µl

Preis
SFr. 512.00
Einloggen für Verfügbarkeit
1 * 100 µl
Product Details & Documents

Type: Primary
Antigen: CCR8 (chemokine (C-C motif) receptor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Technische Daten
  • Größe:
    100 µl
  • Western blot:
    Yes
  • Antikörper-Typ:
    Primär
  • Konjugation:
    Unconjugated
  • Reinigung:
    purified by affinity chromatography
  • Transporttemperatur:
    Ice
  • Reaktivität:
    Human|Rat|Mouse
  • Konzentration:
    1 mg/ml
  • Cat. No.:
    BIRBORB107482-100
  • Antigen-Name:
    chemokine (C-C motif) receptor 8
  • Lagertemperatur:
    Cool
  • Antigen-Symbol:
    CCR8
  • Klonalität:
    Polyklonal
  • Isotyp:
    IgG
  • Wirt:
    Rabbit
  • ELISA:
    Yes
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..