Order Entry
Switzerland
ContactUsLinkComponent
Anti-SART1 Rabbit Polyclonal Antibody
  BIRBORB41664-100
 :  Biorbyt
Anti-SART1 Rabbit Polyclonal Antibody
  BIRBORB41664-100
 :  Biorbyt

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Squamous cell carcinoma antigen recognized by T cells 1
  • Antigen-Symbol:
    SART1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Aminosäurezahl:
    533 to 762
  • Lagerungspuffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Lagertemperatur:
    Cool
  • Konzentration:
    1 mg/ml
  • Transporttemperatur:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 533 and 762 of human SART1.
  • Reinigung:
    purified by antigen affinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Western Blot: 1:500-1:3000

Type: Primary
Antigen: SART1 (squamous cell carcinoma antigen recognized by T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human