Anti-ATM Rabbit polyclonal antibody
BIRBORB14843-100
: Biorbyt
- Antikörper-Typ:Primär
- Antigen-Name:Ataxia Telangiectasia Mutated
- Antigen-Symbol:ATM
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser1981
- Lagerungspuffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:370 kDa
- Lagertemperatur:Cool
- Konzentration:0,5 mg/ml
- Transporttemperatur:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human ATM around the phosphorylation site of Ser1981.
- Reinigung:purified by affinity chromatography
- Größe:100 µg
- VE:100 µG
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat