Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-STAT6 Rabbit Polyclonal Antibody
Catalog # BIRBORB14464-100
Anbieter:  Biorbyt
undefined
Anti-STAT6 Rabbit Polyclonal Antibody
Catalog # BIRBORB14464-100
Anbieter:  Biorbyt

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    signal transducer and activator of transcription 6, interleukin-4 induced
  • Antigen-Symbol:
    STAT6
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Lagerungspuffer:
    PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Lagertemperatur:
    Cool
  • Konzentration:
    1 mg/ml
  • Transporttemperatur:
    Ice
  • Immunogen:
    The antiserum was produced against synthesized non-phosphopeptide derived from human STAT6 around the phosphorylation site of threonine 645 (P -A-TP-I-K).
  • Reinigung:
    purified by affinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

Technische Daten

Über diesen Artikel

Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STAT6 (signal transducer and activator of transcription 6, interleukin-4 induced)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human