Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-MTA1 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-MTA1 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB11044-100
  • BIRBORB11044-200
  • Antigen-Symbol:
  • MTA1
  • MTA1
  • Antigen-Name:
  • Metastasis associated 1
  • Metastasis associated 1
  • Antikörper-Typ:
  • Primär
  • Primär
  • Klonalität:
  • Polyklonal
  • Polyklonal
  • Konjugation:
  • Unconjugated
  • Unconjugated
  • Reaktivität:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Wirt:
  • Rabbit
  • Rabbit
  • Isotyp:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemie:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide
  • Synthetic peptide
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 79 kDa
  • 79 kDa
  • Reinigung:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Konzentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Lagerungspuffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Lagertemperatur:
  • Cool
  • Cool
  • Transporttemperatur:
  • Ice
  • Ice

 

 

Frequently Bought Together