Anti-Protein Z Rabbit Polyclonal Antibody (FITC (Fluorescein))
BIRBORB3715-100
: Biorbyt
- Antikörper-Typ:Primär
- Antigen-Name:Protein Z
- Klonalität:Polyklonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Form:liquid
- Lagerungspuffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Lagertemperatur:Cool
- Konzentration:0,5 mg/ml
- Transporttemperatur:Ice
- Immunogen:Synthetic peptide
- Reinigung:purified by Protein A chromatography
- Größe:100 µg
- VE:100 µG
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: Protein Z
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: