Anti-ZNF274 Rabbit Polyclonal Antibody
AVIVARP39808_P050
- Antikörper-Typ:Primär
- Antigen-Name:zinc finger protein 274
- Antigen-Symbol:ZNF274
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:
- Reaktivität:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitop:middle
- Kreuzadsorption:none
- Gen-ID:10782
- Antigen-Synonyme:HFB101|ZF2|ZKSCAN19|ZSCAN51
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:50 µG
Type: Primary
Antigen: ZNF274 (zinc finger protein 274)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human