Anti-CIDEC Rabbit Polyclonal Antibody
Katalognr. BOSSBS-6796R
Anbieter: Bioss
Direkt zu
Product Details & Documents
Type: Primary
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Dokumente
bs-6796R
Technisches Datenblatt
Technische Daten
- Western blot:Yes
- Antikörper-Typ:Primär
- Konjugation:Unconjugated
- Transporttemperatur:Ice
- Reaktivität:Human|Rat|Mouse
- Kreuzadsorption:none
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Cat. No.:BOSSBS-6796R
- Antigen-Name:Cell death-inducing DFFA-like effector C
- Lagertemperatur:Cool
- Antigen-Synonyme:CIDE-3|FPLD5|CIDE3|FSP27
- Antigen-Symbol:CIDEC
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit
- ELISA:Yes



