Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Ferritin heavy polypeptide
- Antigen-Symbol:FTH1
- Klonalität:Monoklonal
- Klon:3F8
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Isotyp:IgG2a
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:none
- Gen-ID:2495
- Antigen-Synonyme:FHC|PLIF|PIG15|FTH|HFE5|FTHL6
- Lagertemperatur:Store for 1 year at –20 °C from date of shipment
- Transporttemperatur:Ice
- Immunogen:Recombinant human protein purified from E.coli (HIS-Ferritin H chain)
- Reinigung:Ammonium sulfate precipitation
- Größe:100 µl
- VE:0,1 mL
Specifications
About this item
Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (~450 kDa) consisting of 24 subunits of two types, H(heavy) and L(light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.
Western Blot: 1:1,000;
Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human