Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:attractin
- Antigen-Symbol:ATRN
- Klonalität:Monoklonal
- Klon:9H8
- Konjugation:Unconjugated
- Wirt:Mouse
- Immunpräzipitation:Yes
- Isotyp:IgG1
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:none
- Gen-ID:8455
- Antigen-Synonyme:DPPT-L|MGCA
- Lagertemperatur:Store for 1 year at –20 °C from date of shipment
- Transporttemperatur:Ice
- Immunogen:Protein purified from Human plasma
- Reinigung:Ammonium sulfate precipitation
- Größe:100 µl
- VE:0,1 mL
Technische Daten
Über diesen Artikel
Attractin is a serum glycoprotein of 175 kDa and found in both membrane-bound and secreted forms as a result of alternative splicing. Both the secreted and membrane-bound forms of attractin may be involved in the development and maintenance of the central nervous system. Membrane-bound attractin is a co-receptor for Agouti, antagonist of melanocortin-1 receptor. Secreted attractin, expressed by activated T lymphocytes and modulates interactions between T cells and monocytes/macrophages, was examined as a potential marker of immune activity. Attractin may be a component of a pathway for regulated protein turnover that also involves mahogunin, a wide-expressed E3 ubiquitin ligase found at particularly high levels in the brain. Attractin was considered as an extracellular target amenable for the development of obesity-regulating drugs, also.
Western Blot: 1: 1,000; Immunoprecipitation: 1-2 ?l;
Type: Primary
Antigen: ATRN (attractin)
Clonality: Monoclonal
Clone: 9H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human