Ampoule snapper, AccuVac®
Snapper for ampoules
Katalognr. HACH2405200
Anbieter: Hach
Direkt zu
Product Details & Documents
This snapper is used for opening ampoules immersed in samples.
- Avoids sample contact
- Used with colorimeters and spectrophotometers
Technische Daten
- Bezeichnung:Snapper for ampoules
- Seq:01