MF Endo broth
Catalog # 516-9079
Supplier: Cytiva (Formerly Pall Lab)
CAS Number:
Specifications
- VE:1
- Normen:-
- Bezeichnung:MF-Endo Broth, Total Coliforms
- Seq:02
- Füllung:100 ml
Specifications
About this item
M-Endo broth is used for the estimation of coliform bacteria in water samples using the membrane filtration technique. It is possible to remove bacteria from fluids by passing them through filters with such small pore size that bacteria are arrested. This filtration technique enables fairly large volumes of water to pass rapidly under pressure, but prevents the passage of any bacteria present. These nutrients are retained on the surface of the membrane which is then brought into contact with suitable liquid nutrients. These diffuse upwards through the pores thereby inducing the organisms to grow as surface colonies which can be counted.
- Glass bottle
- Lactose non-fermenters form colourless colonies