ContactUsLinkComponent
CytoScan™ LDH Cytotoxicity Assay kit
Katalognr. 786-210
Anbieter:  G-Biosciences
CAS-Nummer:  
undefined
CytoScan™ LDH Cytotoxicity Assay kit
Katalognr. 786-210
Anbieter:  G-Biosciences
CAS-Nummer:  
Technische Daten
  • VE:
    1.000 Assays
  • Bezeichnung:
    CytoScan™-LDH Cytotoxicity Kit
  • Tests pro Kit:
    1000 assays
Technische Daten
Über diesen Artikel
CytoScan™ LDH Cytotoxicity assay quantitatively measures the cytosolic, lactate dehydrogenase (LDH) enzyme, which is released from damaged cells. The released LDH is measured with a coupled enzymatic reaction that results in the conversion of a tetrazolium salt (iodonitrotetrazolium; (INT)) into a red color formazan by diaphorase. The resulting formazan is measured at 490 nm.
  • Colorimetric assay
  • Quantitatively measures LDH release
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening