AuftragsverfolgungKontakt
Immobilised Trypsin
Immobilised Trypsin
Trypsin, immobilised, 1
Katalognr. 786-792
Anbieter:   G-Biosciences
Immobilised Trypsin
Katalognr. 786-792
Anbieter:  G-Biosciences
CAS Nummer:  
Immobilised Trypsin
Immobilised Trypsin
Trypsin, immobilised, 1
CAS Nummer:  
Preis
151.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Trypsin is a serine endopeptidase that specifically cleaves peptide bonds on the carboxy side of s-aminoethyl cysteine, arginine and lysine residues and typically there is little or no cleavage at arginyl-proline and lysyl-proline bonds. The distribution of these residues in proteins allows trypsin digestion to produce peptides that are readily identified by mass spectrometry.
Immobilised Trypsin is TPCK treated trypsin immobilised on 4% agarose that eliminates the contamination of protein digests by the trypsin. The immobilised trypsin is readily removed by separating the agarose from the digestion solution.
Documents
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    Trypsin, immobilised