Magnetic stirring bars
38,1 mm, 7,9 mm, Stirring in extremely small containers
Katalognr. HACH1076416
Anbieter: Hach
Direkt zu
Product Details & Documents
Ideal for stirring in extremely small containers.
Technische Daten
- Ø:7,9 mm
- Für:Stirring in extremely small containers
- Länge:38,1 mm