Reference electrode, Dionex™
pH reference electrode, Ion chromatography (carbohydrate analyses/amino acid analyses)
Catalog # DION044198
Marketsource
Supplier: DIONEX
Jump to:
Product Details & Documents
Detector pH-Ag/AgCl reference electrode which can be used for carbohydrate and amino acid analysis with the Thermo Scientific™ Dionex™ ED electrochemical detector pH reference electrode.
Specifications
- Für:Ion chromatography (carbohydrate analyses/amino acid analyses)
- Bezeichnung:pH reference electrode
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.