PCR tube rack with cover, SP Bel-Art
Catalog # BELAF189020005
Supplier: Bel-Art Products, a Part of SP
Specifications
- Farbe:Gemischt
- Bezeichnung:96-well PCR storage rack with cover
- Seq:03
- VE:5
Specifications
About this item
PP, with translucent PP cover.
Designed for preparing and storing 0,2 ml thin-walled PCR tubes and strip tubes before or after PCR sample processing applications. The compact rack features 96 wells that keep tubes securely upright in an 8×12 array. An alphanumeric grid assists in easy sample identification.
W×D×H: 127×89×34 mm
Ordering information: The Assortment pack includes one of each colour; flourescent blue, green, pink, yellow and orange.