AuftragsverfolgungKontakt
Urine containers, SpecTainer™ | Avantor
Urine containers, SpecTainer™
Sterile, 120 ml
Katalognr. SIMPC566-120CYS
Anbieter:   Simport Scientific
Urine containers, SpecTainer™
Katalognr. SIMPC566-120CYS
Anbieter:  Simport Scientific
Urine containers, SpecTainer™
Urine containers, SpecTainer™
Sterile, 120 ml
Preis
SFr. 183.00
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents
PP urine container with tamper evident PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Tamper evident sealing ring for better sample protection
  • Ridges around base offer a better grip during opening and closing
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer™ I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen. For colour coding purposes, place a Capinsert™ on top of closure (see Series T345). Ten different colours are available.
Dokumente
Technische Daten
  • Ausführung:
    Sterile
  • Kapazität:
    120 ml
  • Seq:
    06