AuftragsverfolgungKontakt
HPLC columns, MetaSil® Basic
HPLC columns, MetaSil® Basic
Supplier:   VARIAN
HPLC columns, MetaSil® Basic
Supplier:  VARIAN
Certificates
HPLC columns, MetaSil® Basic
4 Options Available Below
Product Details & Documents
MetaSil® Basic columns tends to retain polar compounds better than standard phases, often showing improved retention for compounds that elute in the void volume on standard alkyl phases. These columns are recommended for use at pH 2 to 7,5.
  • Excellent peak shape for basic analytes
  • Exceptionally high surface coverage
  • Improved polar compound retention
  • Enhanced selectivity
Product Family Options
Product Information
  • Säulenform
    Porengröße
    Partikelgröße
    Ø int.
    Länge
    Availability
    Price
  • Analytisch
    100 Å
    5 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Analytisch
    100 Å
    3 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Analytisch
    100 Å
    3 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Guard columns
Product Information
  • Säulenform
    Porengröße
    Partikelgröße
    Ø int.
    Länge
    Availability
    Price
  • Vorsäule
    100 Å
    3 µm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST