GC syringes for Thermo Scientific instruments
Catalog # 555-8416
Supplier: Thermo Fisher Scientific
Specifications
- Für:TriPlus, AS2000
- Gauge:26
- Typ:LVOC
- Spitzentyp:Cone
- Volumen:250 µl
- Bezeichnung:Removable needle, gas tight
- Länge:80 mm
- Seq:04
- Environmentally Preferable:
- VE:1