ContactUsLinkComponent
Microinjectors, Nanoject II™
Microinjectors, Nanoject II™
Katalognr. DRUM3-000-206A
Supplier:  DRUMMOND
Microinjectors, Nanoject II™
Katalognr. DRUM3-000-206A
Supplier:  DRUMMOND
Specifications
  • Bezeichnung:
    Nanoject II™, 220 V, UK-plug
  • Seq:
    02
  • VE:
    1
Specifications
About this item
The Nanoject II™ microinjector is specifically designed to perform ultra-delicate nanolitre injection procedures into oocytes, embryos, and tissues.
  • Automated microprocessor controlled microinjection pipette delivering precise nanolitre volumes accurately
  • Remote control and non rotating plunger eliminates vibration, allowing precise injection minimising possible cell rupture
  • Separate foot switch available to allow hands-free operation
  • Can be mounted on most micromanipulators
  • Supplied with sufficient borosilicate glass capillaries for preparing 400 injection tips, standard softening point 780 °C