Biocide, sterile, ready to use, Klercide™ Sporicidal Active Chlorine
Supplier: ECOLAB CONTAMINATION CONTROL
Certificates
About this item
Klercide™ Sporicidal Active Chlorine is an effective sporicide based on active chlorine, derived from a sterile sodium hypochlorite solution. The ready to use version is 0,2 µm filtered, filled and double bagged in a Grade C cleanroom before gamma irradiation to a validated procedure.
- Suitable for class A/B/C/D cleanrooms
- Fully compatible and ideal for a rotational regime
- SteriShield Delivery System™ for 1 litre spray bottle
Certifications: Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697; Sporicidal EN 13704.
Caution: Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.
Frequently Bought Together