Bosshead, multi-purpose
Bosshead, square, with M8 brass thumb bolt, 140 g, 16 mm, Aluminiumlegierung
Katalognr. JUCHVL04825
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Technische Daten
- Spannweite:16 mm
- Bezeichnung:Bosshead, square, with M8 brass thumb bolt, 140 g
- Material:Aluminiumlegierung